missing translation for 'onlineSavingsMsg'
Learn More

CAY10471 Racemate, MedChemExpress

Product Code. 16418487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16418487

Brand: Medchem Express HY13706

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CAY10471 Racemate (TM30089 Racemate) is a potent and highly selective prostaglandin D2 receptor CRTH2 antagonist, with a Ki of 0.6 nM for hCRTH2, selective over human thromboxane A2 receptor TP (Ki, >10000 nM) or PGD2 receptor DP (Ki, 1200 nM). CAY10471 Racemate also has effect on mouse and rat orthologs of CRTH2.

CAY10471 Racemate (TM30089 Racemate) is a potent and highly selective prostaglandin D2 receptor CRTH2 antagonist, with a Ki of 0.6 nM for hCRTH2, selective over human thromboxane A2 receptor TP (Ki, >10000 nM) or PGD2 receptor DP (Ki, 1200 nM). CAY10471 Racemate also has effect on mouse and rat orthologs of CRTH2.

Specifications

Chemical Name or Material CAY10471 Racemate
CAS 844639-57-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H21FN2O4S
Quantity 5 mg
Synonym TM30089 Racemate
Solubility Information DMSO : 150 mg/mL (360.17 mM; Need ultrasonic)
SMILES O=C(O)CN1C2=C(C3=C1C=CC=C3)CC(N(S(=O)(C4=CC=C(F)C=C4)=O)C)CC2
Molecular Weight (g/mol) 416.47
Formula Weight 416.47
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.