missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98712-50ul
This item is not returnable.
View return policy
Description
Caveolin-3 Polyclonal antibody specifically detects Caveolin-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Caveolin-3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| caveolin 3, caveolin-3, LGMD1CMGC126100, LQT9MGC126101, M-caveolin, MGC126129, VIP21, VIP-21 | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the Human Caveolin-3. | |
| 50 μg | |
| Endocrinology, Signal Transduction | |
| 859 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction