missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caveolin-2 Polyclonal antibody specifically detects Caveolin-2 in Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Dilution | Western Blot, Immunoprecipitation |
| Gene Alias | CAV, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, MGC12294 |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Research Discipline | Caveolae Markers, Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?