missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-1 Antibody (2A7), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Caveolin-1 |
|---|---|
| Clone | 2A7 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043748
|
Novus Biologicals
NBP3-26695-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043112
|
Novus Biologicals
NBP3-26695-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISATekniske data
| Caveolin-1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
| A synthesized peptide derived from Human Caveolin-1 [UniProt Q03135] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2A7 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 857 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel