missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cav3.2 Antibody (S55/10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP1-22444
This item is not returnable.
View return policy
Description
Cav3.2 Monoclonal specifically detects Cav3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.
Specifications
| Cav3.2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.1-1.0 ug/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| CACNA1HB, calcium channel, voltage-dependent, T type, alpha 1H subunit, calcium channel, voltage-dependent, T type, alpha 1Hb subunit, Cav3.2, ECA6, EIG6, FLJ90484, Low-voltage-activated calcium channel alpha1 3.2 subunit, low-voltage-activated calcium channel alpha13.2 subunit, voltage dependent t-type calcium channel alpha-1H subunit, voltage-dependent T-type calcium channel subunit alpha-1H, voltage-gated calcium channel alpha subunit Cav3.2, voltage-gated calcium channel alpha subunit CavT.2, Voltage-gated calcium channel subunit alpha Cav3.2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8912 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| S55/10 | |
| Unconjugated | |
| O95180 | |
| CACNA1H | |
| Fusion protein amino acids 1019-1293 (II-III loop) of human Cav3.2 | |
| 0.1 mg | |
| Primary | |
| Detects approx 260 kDa. No cross-reactivity against Cav1.3. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction