missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cav2.2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£385.00 - £537.00
Specifications
| Antigen | Cav2.2 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18312405
|
Novus Biologicals
NBP3-17353-25UL |
25 μg |
£385.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18388706
|
Novus Biologicals
NBP3-17353-100UL |
100 μg |
£537.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cav2.2 Polyclonal antibody specifically detects Cav2.2 in Human samples. It is validated for ImmunofluorescenceSpecifications
| Cav2.2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| PBS, pH 7.2, 40% glycerol | |
| 774 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BIII, Brain calcium channel III, CACNL1A5calcium channel, L type, alpha-1 polypeptide, CACNN, calcium channel alpha12.2 subunit, calcium channel, N type, calcium channel, voltage-dependent, alpha 1B subunit, N type, calcium channel, voltage-dependent, L type, alpha 1B subunit, calcium channel, voltage-dependent, N type, alpha 1B subunit, Cav2.2, Cav2.2 voltage-gated Ca2+ channel, L type, alpha-1 polypeptide isoform 5, voltage-dependent N-type calcium channel subunit alpha-1B, Voltage-gated calcium channel subunit alpha Cav2.2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ALMIFDFYKQNKTTRDQMQQAPGGLSQMGPVSLFHPLKATLEQTQPAVLRGARVFLRQKSSTSLSNGGAIQNQESGIKESVSWGTQRTQDAPHE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title