missing translation for 'onlineSavingsMsg'
Learn More

Cav2.2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18356972 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356972 100 μg 100µg
18302592 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18356972 Supplier Novus Biologicals Supplier No. NBP311889100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cav2.2 Polyclonal antibody specifically detects Cav2.2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav2.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 5 - 10 ug/ml
Formulation PBS
Gene Alias BIII, Brain calcium channel III, CACNL1A5calcium channel, L type, alpha-1 polypeptide, CACNN, calcium channel alpha12.2 subunit, calcium channel, N type, calcium channel, voltage-dependent, alpha 1B subunit, N type, calcium channel, voltage-dependent, L type, alpha 1B subunit, calcium channel, voltage-dependent, N type, alpha 1B subunit, Cav2.2, Cav2.2 voltage-gated Ca2+ channel, L type, alpha-1 polypeptide isoform 5, voltage-dependent N-type calcium channel subunit alpha-1B, Voltage-gated calcium channel subunit alpha Cav2.2
Host Species Rabbit
Immunogen Synthetic peptide derived from the rat alpha1B calcium channel conjugated to KLH (Uniprot: Q00975)
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 774
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.