missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cav2.2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92336-0.02ml
This item is not returnable.
View return policy
Description
Cav2.2 Polyclonal antibody specifically detects Cav2.2 in Human samples. It is validated for Western Blot
Specifications
| Cav2.2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BIII, Brain calcium channel III, CACNL1A5calcium channel, L type, alpha-1 polypeptide, CACNN, calcium channel alpha12.2 subunit, calcium channel, N type, calcium channel, voltage-dependent, alpha 1B subunit, N type, calcium channel, voltage-dependent, L type, alpha 1B subunit, calcium channel, voltage-dependent, N type, alpha 1B subunit, Cav2.2, Cav2.2 voltage-gated Ca2+ channel, L type, alpha-1 polypeptide isoform 5, voltage-dependent N-type calcium channel subunit alpha-1B, Voltage-gated calcium channel subunit alpha Cav2.2 | |
| A synthetic peptide corresponding to a sequence within amino acids 2200 to the C-terminus of human CACNA1B (NP_000709.1). SITYKTANSSPIHFAGAQTSLPAFSPGRLSRGLSEHNALLQRDPLSQPLAPGSRIGSDPYLGQRLDSEASVHALPEDTLTFEEAVATNSGRSSRTSYVSSLTSQSHPLRRVPNGYHCTLGLSSGGRARHSYHHPDQDHWC | |
| 0.02 mL | |
| Neuroscience, Signal Transduction | |
| 774 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction