missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaV2.1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549496
This item is not returnable.
View return policy
Description
CaV2.1 Polyclonal Antibody for Western Blot
Voltage-dependent calcium channels mediate the entry of calcium ions into excitable cells, and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, and gene expression. Calcium channels are multisubunit complexes composed of alpha-1, beta, alpha-2/delta, and gamma subunits. The channel activity is directed by the pore-forming alpha-1 subunit, whereas, the others act as auxiliary subunits regulating this activity. The distinctive properties of the calcium channel types are related primarily to the expression of a variety of alpha-1 isoforms, alpha-1A, B, C, D, E, and S. This gene encodes the alpha-1A subunit, which is predominantly expressed in neuronal tissue. Mutations in this gene are associated with 2 neurologic disorders, familial hemiplegic migraine and episodic ataxia 2. This gene also exhibits polymorphic variation due to n-repeats. Multiple transcript variants encoding different isoforms have been found for this gene. In one set of transcript variants, the n-repeats occur in the 3 UTR, and are not associated with any disease. But in another set of variants, an insertion extends the coding region to include the n-repeats which encode a polyglutamine tract. Expansion of the n-repeats from the normal 4-16 to 21-28 in the coding region is associated with spinocerebellar ataxia 6.
Specifications
| CaV2.1 | |
| Polyclonal | |
| Unconjugated | |
| CACNA1A | |
| alpha1A; APCA; BccA1; BI; brain calcium channel 1; brain calcium channel I; brain class A; Caca1a; Cach4; CACN3; CACNA1A; Cacnl1a4; calcium channel alpha 1A; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 4; Calcium channel, L type, alpha-1 polypeptide, isoform 4; calcium channel, voltage-dependent, alpha 1A subunit; calcium channel, voltage-dependent, P/Q type, alpha 1A subunit; calcium voltage-gated channel subunit alpha1 A; Cav2.1; Ccha1a; EA2; fetal brain Ca2+ voltage-gated channel alpha1A pore-forming subunit; FHM; HPCA; la; leaner; MHP; MHP1; nmf352; Rat brain class A; rbA-1; RBA-I; rkr; rocker; SCA6; tg; tottering; voltage-dependent P/Q-type calcium channel subunit alpha-1A; Voltage-gated calcium channel subunit alpha Cav2.1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O00555 | |
| CACNA1A | |
| KLH conjugated synthetic peptide between 898-932 amino acids from the Central region of human CACNA1A. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction