missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cav1.4 Antibody (1H6), Novus Biologicals™

Product Code. 18345897 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345897 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345897 Supplier Novus Biologicals™ Supplier No. H00000778M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cav1.4 Monoclonal antibody specifically detects Cav1.4 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav1.4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1H6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005174
Gene Alias AIED, CACNAF1, calcium channel, voltage-dependent, L type, alpha 1F subunit, Cav1.4, Cav1.4alpha1, COD4, CORDX, CORDX3CSNB2, CSNB2AAland island eye disease (Forsius-Eriksson ocular albinism, ocular albinismtype 2), CSNBX2, JM8, JMC8, OA2COD3, voltage-dependent L-type calcium channel subunit alpha-1F, Voltage-gated calcium channel subunit alpha Cav1.4
Host Species Mouse
Immunogen CACNA1F (NP_005174, 1878 a.a. ∽ 1977 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LHVPGTHSDPSHGKRGSADSLVEAVLISEGLGLFARDPRFVALAKQEIADACRLTLDEMDNAASDLLAQGTSSLYSDEESILSRFDEEDLGDEMACVHAL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 778
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.