Learn More
Cav1.3 Antibody (S48), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-12893-0.025mg
 View more versions of this product
Product Code. 18782994
  • £176.00 / 0.03mg
Description
Cav1.3 Monoclonal specifically detects Cav1.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cav1.3
Monoclonal
1 mg/mL
Western Blot 1 - 10 μg/mL, Immunohistochemistry 0.1 - 1 μg/mL, Immunocytochemistry/Immunofluorescence 1 - 10 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Microarray
CACNA1D
Fusion protein amino acids 859-875 of rat Cav1.3
0.025 mg
Primary
Detects approx 250 kDa. No cross-reactivity against Cav1.2.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a κ
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay
S48
Unconjugated
CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3
Mouse
Protein G purified
RUO
776
Human, Mouse, Rat
Purified