missing translation for 'onlineSavingsMsg'
Learn More

Cav1.3 Antibody (S48), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18427274 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427274 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427274 Supplier Novus Biologicals Supplier No. NBP212893JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cav1.3 Monoclonal antibody specifically detects Cav1.3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav1.3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S48
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3
Host Species Mouse
Immunogen Fusion protein amino acids 859-875 of rat Cav1.3
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 776
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.