missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaV1.2 Monoclonal Antibody (S57), PerCP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545389
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45389 was sufficient for detection of Cav1.2 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 240kDa (varies with cell background due to glycosylation).
CACNA1C encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by CACNA1C binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins.
Specifications
| CaV1.2 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 2mM EDTA and no preservative; pH 7.4 | |
| P22002, Q01815, Q13936 | |
| CACNA1C | |
| Fusion protein amino acids 1507-1733 (intracellular carboxyl terminus) of rabbit Cav1.2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Hamster | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry, Microarray | |
| S57 | |
| PerCP | |
| CACNA1C | |
| brain class C; CACB; CaCB receptor; CACB-receptor; CACH2; CACH2C; CACH3; Cacn2; CACN4; CACNA 1D; Cacna1c; CACNL1A1; CACNL1A2; calcium channel; calcium channel voltage-dependent alpha1c subunit; calcium channel, cardic dihydropyridine-sensitive, alpha-1 subunit; calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle; calcium channel, voltage-dependent, alpha 1C subunit; calcium channel, voltage-dependent, L type, alpha 1C subunit; calcium voltage-gated channel subunit alpha1 C; cardiac L-type calcium channel; Cav1.2; CCHL1A1; D930026N18Rik; DHP receptor; DHPR, alpha-1 subunit; I79_006562; long-NT; LQT8; LTCC; L-type calcium channel alpha-1 subunit; MBC; MELC-CC; MGC120730; Mouse brain class C; neuronal voltage-gated calcium channel alpha 1C subunit; Rat brain class C; RATIVS302; RBC; skeletal muscle-specific calcium channel; smooth muscle calcium channel blocker; Smooth muscle calcium channel blocker receptor; TS; unnamed protein product; voltage-dependent L-type Ca2+ channel alpha 1 subunit; Voltage-dependent L-type calcium channel subunit alpha-1C; Voltage-gated calcium channel subunit alpha Cav1.2; voltage-gated L-type calcium channel Cav1.2 alpha 1 subunit, splice variant 10* | |
| Mouse | |
| Protein G | |
| RUO | |
| 100754788, 12288, 24239, 775 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction