missing translation for 'onlineSavingsMsg'
Learn More

Cav1.2 Antibody (S57), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18430024 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430024 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430024 Supplier Novus Biologicals Supplier No. NBP122439AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cav1.2 Monoclonal antibody specifically detects Cav1.2 in Human, Mouse, Rat, Hamster, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav1.2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S57
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias alpha-1 subunit, calcium channel, voltage-dependent, L type, alpha 1C subunit, cardic dihydropyridine-sensitive, alpha-1 subunit, L type, alpha 1 polypeptide, isoform 1, cardic muscle, L type, alpha-1 polypeptide, isoform 1, cardiac muscle, splice variant 10*
Host Species Mouse
Immunogen Fusion protein amino acids 1507-1733 (intracellular carboxyl terminus) of rabbit Cav1.2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cognition and Behavior, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 775
Target Species Human, Mouse, Rat, Hamster, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.