missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Cathode Buffer Container (CBC) for 3500 Series Systems for Protein Quality Analysis
Shop All Applied Biosystems Products
Click to view available options
Quantity:
4 Containers
Unit Size:
Pack of 4
Specifications
Specifications
| Content And Storage | Contains 4 Cathode Buffer containers. Store at 2–8°C until ready to use. Refer to the expiration date on the label. |
| For Use With (Equipment) | 3500 Genetic Analyzer for Protein Quality Analysis, 3500xL Genetic Analyzer for Protein Quality Analysis |
| For Use With (Application) | Sequencing |
| Product Type | Cathode Buffer Container |
| Quantity | 4 Containers |
| Shipping Condition | Wet Ice |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction