missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Cathode Buffer Container (CBC) for 3500 Series Systems for Protein Quality Analysis

Product Code. 15889876 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Containers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15889876 4 Containers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889876 Supplier Applied Biosystems™ Supplier No. A31279

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Cathode Buffer Container (CBC) contains 1X running buffer to support protein quality analysis applications on the Applied Biosystems™ 3500 Series genetic analyzers for protein quality analysis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 4 Cathode Buffer containers.
Store at 2–8°C until ready to use. Refer to the expiration date on the label.
For Use With (Equipment) 3500 Genetic Analyzer for Protein Quality Analysis, 3500xL Genetic Analyzer for Protein Quality Analysis
For Use With (Application) Sequencing
Product Type Cathode Buffer Container
Quantity 4 Containers
Shipping Condition Wet Ice

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.