missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Cathode Buffer Container (CBC), for 3500/SeqStudio™ Flex
Shop All Applied Biosystems Products
missing translation for 'orderingAttributeHoverText'
Quantity:
4 pack
missing translation for 'unitSize'
Pack of 4
Description
The cathode buffer container (CBC) contains 1X running buffer to support all electrophoresis applications on Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers. The container has two separate compartments, the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections.
The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the CBC is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.
The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the CBC is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.
Order Info
4 Pack
Specifications
Specifications
| Content And Storage | Contains 4 packs of cathode buffer container. Store the Cathode Buffer Container at 2-8°C until ready to use. Refer to the expiration date on the label. |
| For Use With (Equipment) | 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers |
| Type | Cathode Buffer Container |
| Green Features | Sustainable packaging |
| Quantity | 4 pack |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction