missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Cathode Buffer Container Dx

Product Code. 16305332 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Ea.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16305332 4 Ea. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16305332 Supplier Applied Biosystems™ Supplier No. 4408258

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers.

The Cathode Buffer Container Dx is for use with Applied Biosystems Dx genetic analyzers.

The Cathode Buffer Container Dx contains 1X running buffer to support electrophoresis applications on Applied Biosystems Dx genetic analyzers. The Cathode Buffer Container Dx has two separate compartments: the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections. The Cathode Buffer Container Dx is a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the Cathode Buffer Container Dx is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers. The Septa Cathode Buffer Container Dx (sold separately) is used for sealing the Cathode Buffer Container Dx container.

This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.

The cathode buffer container (CBC) contains 1x running buffer to support all electrophoresis applications on the 3500 Dx series Genetic Analyzers.

Order Info

4 Pack

TRUSTED_SUSTAINABILITY

Specifications

Starting Material DNA, RNA
Content And Storage Each package contains 4 buffer containers.
Store at 2 - 8°C.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product Type Cathode Buffer Container
Quantity 4 Ea.
Shipping Condition Wet Ice

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.