missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin Z Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547048
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 40% cross-reactivity with recombinant human Cathepsin X/Z/P is observed and less than 2% cross-reactivity with recombinant mouse Cathepsin B, C, D, H, and L is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis.
Specifications
| Cathepsin Z | |
| Polyclonal | |
| Unconjugated | |
| Ctsz | |
| AI787083; AU019819; carboxypeptidase LB; cathepsin B2; cathepsin IV; cathepsin P; Cathepsin X; cathepsin Y; cathepsin Z; cathepsin Z1; CTSX; CTSZ; cysteine-type carboxypeptidase; D2Wsu143e; lysosomal carboxypeptidase B; preprocathepsin P | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 64138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Neutralization, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9WUU7 | |
| Ctsz | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin X/Z/P Ala23-Val306. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction