missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin X/Z/P Antibody (6G11), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18401146 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401146 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401146 Supplier Novus Biologicals Supplier No. NBP231211MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cathepsin X/Z/P Monoclonal antibody specifically detects Cathepsin X/Z/P in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin X/Z/P
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6G11
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CTSX
Host Species Mouse
Immunogen Recombinant human Cathepsin X/Z/P (62-303aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Epitope Tags
Primary or Secondary Primary
Gene ID (Entrez) 1522
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.