missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin X/Z/P Antibody (6G11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-31211CL1
This item is not returnable.
View return policy
Description
Cathepsin X/Z/P Monoclonal antibody specifically detects Cathepsin X/Z/P in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Cathepsin X/Z/P | |
| Monoclonal | |
| CoraFluor 1 | |
| CTSX | |
| Recombinant human Cathepsin X/Z/P (62-303aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Epitope Tags | |
| 1522 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 6G11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction