missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin X/Z/P Antibody (6G11), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31211AF594
This item is not returnable.
View return policy
Description
Cathepsin X/Z/P Monoclonal specifically detects Cathepsin X/Z/P in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Cathepsin X/Z/P | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 6G11 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| CTSZ | |
| Recombinant human CTSZ (62-303aa) purified from E. coli | |
| Cancer, Epitope Tags | |
| 1522 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction