missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin X/Z/P Antibody (009), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90411AF532
This item is not returnable.
View return policy
Description
Cathepsin X/Z/P Monoclonal specifically detects Cathepsin X/Z/P in Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Cathepsin X/Z/P | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin X/Z/P (NP_071720.1; Met 1-Val 306). | |
| 0.1 mL | |
| Cancer, Epitope Tags | |
| 1522 | |
| Mouse | |
| Purified |
| ELISA, Immunofluorescence | |
| 9 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CTSZ | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction