missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin W Antibody (4A8), Novus Biologicals™

Product Code. 18335308 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335308 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335308 Supplier Novus Biologicals Supplier No. H00001521M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cathepsin W Monoclonal antibody specifically detects Cathepsin W in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin W
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4A8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH48255
Gene Alias cathepsin W, cathepsin W (lymphopain), EC 3.4.22.-, Lymphopain, LYPN
Host Species Mouse
Immunogen CTSW (AAH48255, 22 a.a. ~ 376 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IRGPLRAQDLGPQPLELKEAFKLFQIQFNRSYLSPEEHAHRLDIFAHNLAQAQRLQEEDLGTAEFGVTPFSDLTEEEFGQLYGYRRAAGGVPSMGREIRSEEPEESVPFSCDWRKVAGAISPIKDQKNCNCCWAMAAAGNIETLWRISFWDFVDVSVQELLDCGRCGDGCHGGFVWDAFITVLNNSGLASEKDYPFQGKVRAHRCHPKKYQKVAWIQDFIMLQNNEHRIAQYLATYGPITVTINMKPLQLYRKGVIKATPTTCDPQLVDHSVLLVGFGSVKSEEGIWAETVSSQSQPQPPHPTPYWILKNSWGAQWGEKGYFRLHRGSNTCGITKFPLTARVQKPDMKPRVSCPP
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1521
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.