missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin V Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18442-1-AP-20UL
This item is not returnable.
View return policy
Description
Cathepsin V (CTSU, CTSV, CATL2, cathepsin U, cathepsin L2) is a lysosomal cysteine proteinase that influences corneal physiology, and mediates degradation of invariant chain in human thymus. Cathepsin V is present in corneal epithelium, activated macrophages, and colorectal and breast carcinomas. A 1.8-kb mRNA to Cathepsin V is present in normal human thymus and testis. Cathepsin V is a member of the peptidase C1 family. Cysteine proteinases are synthesized as preproenzymes, which are processed to the corresponding proenzymes. The proenzymes are either targeted to the lysosome or continue along the cellular secretory route.Specifications
| Cathepsin V | |
| Polyclonal | |
| Unconjugated | |
| CTSV | |
| cathepsin L2, Cathepsin U, Cathepsin V, CATL2, CTSL2, CTSU, CTSV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1515 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60911 | |
| CTSV | |
| Cathepsin V Fusion Protein Ag13284 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur