missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin S Recombinant Rabbit Monoclonal Antibody (HL2462)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550540
This item is not returnable.
View return policy
Description
Cathepsin S Recombinant Monoclonal Antibody for Western Blot, IHC (P)
The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.
Specifications
| Cathepsin S | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P25774 | |
| CTSS | |
| Recombinant fragment of human Cathepsin S. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2462 | |
| Unconjugated | |
| CTSS | |
| Cat S; cathepsin S; Cats; CTSS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction