missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin S Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580521
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Cathepsin S/CTSS.
The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.
Specifications
| Cathepsin S | |
| Polyclonal | |
| Unconjugated | |
| CTSS | |
| Cat S; cathepsin S; Cats; CTSS | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| P25774 | |
| CTSS | |
| Recombinant Human Cathepsin S/CTSS protein (Met1-Ile331). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction