Learn More
Invitrogen™ Cathepsin S Monoclonal Antibody (K29028_13C4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556305
Description
Sequence of this protein is as follows: LPDSVDWREK GCVTEVKYQG SCGACWAFSA VGALEAQLKL KTGKLVSLSA QNLVDCSTEK YGNKGCNGGF MTTAFQYIID NKGIDSDASY PYKAMDQKCQ YDSKYRAATC SKYTELPYGR EDVLKEAVAN KGPVSVGVDA RHPSFFLYRS GVYYEPSCTQ NVNHGVLVVG YGDLNGKEYW LVKNSWGHNF GEEGYIRMAR NKGNHCGIAS FPSYPEI
The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.
Specifications
| Cathepsin S | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P25774 | |
| CTSS | |
| Recombinant human CTSS (immunogen range is 115-331). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| K29028_13C4 | |
| Unconjugated | |
| CTSS | |
| Cat S; cathepsin S; Cats; CTSS | |
| Mouse | |
| Protein A | |
| RUO | |
| 1520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.