missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin S Antibody (SR1393), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | Cathepsin S |
|---|---|
| Clone | SR1393 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621394
|
Novus Biologicals
NBP3-21900-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616664
|
Novus Biologicals
NBP3-21900-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Cathepsin S | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
| A synthesized peptide derived from human Cathepsin S (Uniprot #: P25774) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1393 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1520 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title