missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin S Antibody (054), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90713AF700
This item is not returnable.
View return policy
Description
Cathepsin S Monoclonal specifically detects Cathepsin S in Mouse samples. It is validated for ELISA.
Specifications
| Cathepsin S | |
| Monoclonal | |
| Alexa Fluor 700 | |
| cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin S (AAB94925.1; Met1-Ile340). | |
| 0.1 mL | |
| Immunology | |
| 1520 | |
| Mouse | |
| Purified |
| ELISA | |
| 54 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CTSS Mouse CTSA Mouse CTSB Mouse CTSC Mouse CTSD Mouse CTSE Mouse CTSX | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction