missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cathepsin O Recombinant Mouse Monoclonal Antibody (K06376_14F12)
GREENER_CHOICE

Product Code. 30828543
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828543 100 μL
30825068 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828543 Supplier Invitrogen™ Supplier No. MA564491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: VESAYAIKGK PLEDLSVQQV IDCSYNNYGC NGGSTLNALN WLNKMQVKLV KDSEYPFKAQ NGLCHYFSGS HSGFSIKGYS AYDFSDQEDE Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 37%, Pig - 88%, Cynomolgus monkey - 98%.

The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in cellular metabolism, participating in various events such as peptide biosynthesis and protein degradation. Cathepsin O maintains the structural features characteristic of other cysteine proteinases including the active site cysteine residue that is involved in covalent intermediate formation during peptide hydrolysis. Cathepsin O is an endoproteinase that may be involved in extracellular matrix degradation. Cathepsin O is abundantly expressed in the ovary, kidney and placenta with lower levels found in thymus and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin O
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K06376_14F12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CTSO
Gene Accession No. P43234
Gene Alias A330105D01Rik; A430056C07; AI118514; cathepsin O; Ctso; CTSO1; LOC684529; similar to cathepsin O
Gene Symbols CTSO
Host Species Mouse
Immunogen Recombinant human CTSO (aa 141-230).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1519
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.