missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin O Monoclonal Antibody (180406)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560933
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in cellular metabolism, participating in various events such as peptide biosynthesis and protein degradation. Cathepsin O maintains the structural features characteristic of other cysteine proteinases including the active site cysteine residue that is involved in covalent intermediate formation during peptide hydrolysis. Cathepsin O is an endoproteinase that may be involved in extracellular matrix degradation. Cathepsin O is abundantly expressed in the ovary, kidney and placenta with lower levels found in thymus and skeletal muscle.
Specifications
| Cathepsin O | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P43234 | |
| CTSO | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin O, Asp24-Val321. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 180406 | |
| Unconjugated | |
| CTSO | |
| A330105D01Rik; A430056C07; AI118514; cathepsin O; Ctso; CTSO1; LOC684529; similar to cathepsin O | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1519 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction