missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin L/V/K/H Rabbit anti-Human, Mouse, Rat, Clone: 2Q10F10, Novus Biologicals™

Product Code. 18313294 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18313294 20 μg 20µL
18389026 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18313294 Supplier Novus Biologicals Supplier No. NBP31652420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cathepsin L/V/K/H Monoclonal antibody specifically detects Cathepsin L/V/K/H in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin L/V/K/H
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2Q10F10
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ACC4, ACC-4, ACC5, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin K, cathepsin L1, cathepsin L2, cathepsin L2, preproprotein, Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin U, Cathepsin X, CATL, CATL2, CPSB, CTS02, CTSL1, CTSL2, CTSO, CTSO1, CTSO2, CTSU, Major excreted protein, MEP, N-benzoylarginine-beta-naphthylamide hydrolase, PKND, procathepsin L, PYCD
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Cathepsin L/V/K/H (P07711).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1514
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.