missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin L Antibody (33/2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
£170.00
Specifications
| Antigen | Cathepsin L |
|---|---|
| Clone | 33/2 |
| Dilution | Western Blot 1:200, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Microarray |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Cathepsin L Monoclonal specifically detects Cathepsin L in Human, Mouse, Rat, Mustelid samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Microarray.Specifications
| Cathepsin L | |
| Western Blot 1:200, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Microarray | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 15mM Sodium Azide | |
| cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
| CTSL | |
| IgG1 | |
| Unpurified | |
| Reacts specifically with native and denatured forms of human cathepsin L and procathepsin L. Recognizes an epitope within amino acid residues GYGFEST (265-271 in procathepsin L and 169-175 in the mature cathepsin L molecule). The antibody may be used for the suppression of the malignant growth of myeloma cells. Cross-reactivity is observed with rat cathepsin L (strong), mouse procathepsin L (weak) and mink cathepsin and procathepsin L. Does not cross-react with human cathepsin types B, D, H and S, procathepsins B and D, rat cathepsin B, and mouse procathepsins B and D. |
| 33/2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P07711 | |
| 1514 | |
| Purified full length native protein (Human lung cancer line EPLC 32 M1). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title