missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin L Antibody (221), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18370701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370701 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370701 Supplier Novus Biologicals Supplier No. NBP289528AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cathepsin L Monoclonal specifically detects Cathepsin L in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin L
Applications Western Blot
Classification Monoclonal
Clone 221
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin L (NP_001903.1; Met1-Val333).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1514
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.