missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cathepsin K Substrate (fluorogenic)

For cellular and molecular biology applications

Brand:  Thermo Scientific Chemicals J64082.MA

Code : NEW

Additional Details : CAS Number : 156192-32-4 Weight : 0.00010kg

 View more versions of this product

Product Code. 15446189

  • £104.00 / 10mg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

A sensitive fluorogenic substrate for cathepsin K

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White
A sensitive fluorogenic substrate for cathepsin K
578.65
10mg
Solid
Cathepsin Substrate
-30° to -10°C
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only