missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin K Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11239-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predomitly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing.Specifications
| Cathepsin K | |
| Polyclonal | |
| Unconjugated | |
| CTSK | |
| Cathepsin K, Cathepsin O, Cathepsin O2, Cathepsin X, CTS02, CTSK, CTSO, CTSO1, CTSO2, PKND, PYCD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13038, 1513, 29175 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35186, P43235, P55097 | |
| Ctsk | |
| Cathepsin K Fusion Protein Ag1761 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction