missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin K Antibody (CTSK/2792), PE, Novus Biologicals™

Product Code. 18633920 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18633920 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18633920 Supplier Novus Biologicals Supplier No. NBP279890PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cathepsin K Monoclonal antibody specifically detects Cathepsin K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin K
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone CTSK/2792
Conjugate PE
Formulation PBS
Gene Alias cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD
Host Species Mouse
Immunogen Recombinant fragment of human Cathepsin K protein (around aa 163-274) (exact sequence is proprietary) (Uniprot: P43235)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1513
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.