missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Recombinant Rabbit Monoclonal Antibody (12H15L69), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 703590
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Mouse, Rat, Bovine.
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.Specifications
Cathepsin G | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P08311 | |
CTSG | |
Protein corresponding to human Cathepsin G [aa78-aa196] | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
12H15L69 | |
Unconjugated | |
CTSG | |
CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP | |
Rabbit | |
Protein A | |
RUO | |
1511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |