missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cathepsin G Polyclonal Antibody
GREENER_CHOICE

Product Code. 16559211
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16559211 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16559211 Supplier Invitrogen™ Supplier No. PA596373

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: Mouse liver; Cellular Location: Cell surface.

Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin G
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CTSG
Gene Accession No. P08311, P17977, P28293
Gene Alias CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP
Gene Symbols CTSG
Host Species Rabbit
Immunogen Recombinant protein (or fragment.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13035, 1511, 290257
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.