missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA583878
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GRVSMRRGTD TLREVQLRVQ RDRQCLRIFG SYDPRRQICV GDRRERKAAF K
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.Specifications
Cathepsin G | |
Polyclonal | |
Unconjugated | |
CTSG | |
CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP | |
Rabbit | |
Anitgen affinity chromatography, Antigen affinity chromatography | |
RUO | |
1511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P08311 | |
CTSG | |
Recombinant protein corresponding to Human CTSG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |