missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA579096
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.Specifications
Cathepsin G | |
Polyclonal | |
Unconjugated | |
CTSG | |
CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1511 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5MG BSA and 0.05MG sodium azide | |
P08311 | |
CTSG | |
E. coli-derived human Cathepsin G recombinant protein (Position: R25-E252). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |