missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cathepsin G Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Native Cathepsin G
The Cathepsin G Native Protein is derived from Human neutrophil. The Cathepsin G Native Protein has been validated for the following applications: SDS-Page, Bioactivity.
The Cathepsin G Native Protein is derived from Human neutrophil. The Cathepsin G Native Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Gene ID (Entrez) | 1511 |
| Molecular Weight (g/mol) | 23.5 kDa |
| Quantity | 100 μg |
| Source | Human neutrophil |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Gene Symbol | CTSG |
| Specific Reactivity | Cathepsin G |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction