missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33320-20ul
This item is not returnable.
View return policy
Description
Cathepsin G Monoclonal antibody specifically detects Cathepsin G in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Cathepsin G | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| cathepsin G, CathepsinG, CGCATG, EC 3.4.21, EC 3.4.21.20, MGC23078 | |
| A synthetic peptide corresponding to a sequence within amino acids 156-255 of human Cathepsin G (NP_001902.1).,, Sequence:, TDTLREVQLRVQRDRQCLRIFGSYDPRRQICVGDRRERKAAFKGDSGGPLLCNNVAHGIVSYGKSSGVPPEVFTRVSSFLPWIRTTMRSFKLLDQMETPL | |
| 20 μL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 1511 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction