missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin G Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £379.00
Specifications
| Antigen | Cathepsin G |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698921
|
Novus Biologicals
NBP2-92789-0.02ml |
0.02 mL |
£171.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621192
|
Novus Biologicals
NBP2-92789-0.1ml |
0.1 mL |
£379.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin G Polyclonal antibody specifically detects Cathepsin G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Cathepsin G | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1511 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cathepsin G, CathepsinG, CGCATG, EC 3.4.21, EC 3.4.21.20, MGC23078 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 21-255 of human Cathepsin G (NP_001902.1). IIGGRESRPHSRPYMAYLQIQSPAGQSRCGGFLVREDFVLTAAHCWGSNINVTLGAHNIQRRENTQQHITARRAIRHPQYNQRTIQNDIMLLQLSRRVRRNRNVNPVALPRAQEGLRPGTLCTVAGWGRVSMRRGTDTLREVQLRVQRDRQCLRIFGSYDPRRQICVGDRRERKAAFKGDSGGPLLCNNVAHGIVSYGKSSGVPPEVFTRVSSFLPWIRTTMRSFKLLDQMETPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title