Learn More
Cathepsin D Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547046
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) Cathepsin D is observed and less than 1% cross-reactivity with rhCathepsin A, B, C, L, X, and recombinant mouse (rm) Cathepsin B, C, H, and X is observed. In Western blots, approximately 100% cross-reactivity with rhCathepsin D is observed and less than 1% cross-reactivity with rmCathepsin E is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cathepsin D Polyclonal antibody specifically detects Cathepsin D in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
Cathepsin D | |
Polyclonal | |
Unconjugated | |
Ctsd | |
CatD; Cathepsin D; Cathepsin D heavy chain; Cathepsin D light chain; CathepsinD; CD; Ceroid lipofuscinosis neuronal 10; ceroid-lipofuscinosis, neuronal 10; CLN10; CPSD; CTSD; epididymis secretory sperm binding protein Li 130P; HEL-S-130P; lysosomal aspartyl peptidase; lysosomal aspartyl protease; MGC2311; OTTHUMP00000196039; OTTHUMP00000198692 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13033 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P18242 | |
Ctsd | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin D Ile21-Leu410 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |