missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin D Antibody (2H4H5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £441.00
Specifications
| Antigen | Cathepsin D |
|---|---|
| Clone | 2H4H5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18270271
|
Novus Biologicals
NBP2-52451 |
0.1 mg |
£441.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646816
|
Novus Biologicals
NBP2-52451-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin D Monoclonal specifically detects Cathepsin D in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Cathepsin D | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers | |
| PBS with 0.05% Sodium Azide | |
| 1509 | |
| Purified recombinant fragment of human CTSD (AA: 1-100) expressed in E. Coli. | |
| Primary | |
| 44.6 kDa |
| 2H4H5 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
| CTSD | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title