missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin D Antibody (102), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90432AF700
This item is not returnable.
View return policy
Description
Cathepsin D Monoclonal specifically detects Cathepsin D in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Cathepsin D | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 102 | |
| Western Blot, ELISA | |
| cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin D (NP_034113.1; Met 1-Leu 410). | |
| 0.1 mL | |
| Cancer, Cellular Markers | |
| 1509 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction