Learn More
Invitrogen™ Cathepsin C Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547049
Description
In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Cathepsin C is observed and less than 1% cross-reactivity with recombinant mouse (rm) Cathepsin A, rmCathepsin B, rmCathepsin D, rmCathepsin H, and rmCathepsin X/Z/P is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cathepsin C, known also as dipeptidyl aminopeptidase I (DPPI), is a tetrameric lysosomal cysteine peptidase belonging to the papain family. Cathepsin C is involved in intracellular protein degradation and the processing of protein precursors, where it participates in cell growth, neuraminidase activation, and platelet factor XIII activation. Cathepsin C is largely related to other lysosomal cysteine proteinases, including cathepsin B, H and L. Enzymatically, Cathepsin C is capable of sequentially removing dipeptides from the amino terminus, and it requires halide ions, namely chloride ions, and thiols for complete enzymatic activity. Protein levels of Cathepsin C are detected in a variety of tissues, and it is most highly expressed in spleen, kidney, cytotoxic lymphocytes and myeloid cells, where it localizes to the secretory granule compartment. Cathepsin C is initially synthesized as a proenzyme that is rapidly processed to generate two distinct chains that function together as the mature form of the enzyme.
Specifications
| Cathepsin C | |
| Polyclonal | |
| Unconjugated | |
| CTSC | |
| AI047818; CATC; Cathepsin C; cathepsin J; CPPI; CTSC; Dipeptidyl peptidase 1; Dipeptidyl peptidase 1 exclusion domain chain; Dipeptidyl peptidase 1 heavy chain; Dipeptidyl peptidase 1 light chain; Dipeptidyl peptidase I; Dipeptidyl peptidase I exclusion domain chain; Dipeptidyl peptidase I heavy chain; Dipeptidyl peptidase I light chain; Dipeptidyl transferase; dipeptidyl-peptidase; dipeptidylpeptidase 1; dipeptidyl-peptidase 1; dipeptidyl-peptidase I; DPP1; DPPI; DPP-I; HMS; JP; JPD; PALS; PDON1; PLS | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13032, 25423 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P80067, P97821 | |
| CTSC | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin C/DPPI Asp25-Leu462. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.