missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin C/DPPI Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | Cathepsin C/DPPI |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Cathepsin C/DPPI Polyclonal specifically detects Cathepsin C/DPPI in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cathepsin C/DPPI | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS buffer, 2% sucrose | |
| 1075 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cathepsin CEC 3.4.14.1, Cathepsin J, CPPIHMS, dipeptidyl peptidase 1, Dipeptidyl peptidase I, Dipeptidyl transferase, dipeptidyl-peptidase I, DPP1, DPPI, DPP-I, JP, JPD, PALS, PLS | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human Cathepsin C/DPPI. Peptide sequence VNIAHLKNSQEKYSNRLYKYDHNFVKAINAIQKSWTATTYMEYETLTLGD | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title